Learn More
UBE2E2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303484A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse
UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.Specifications
UBE2E2 | |
Polyclonal | |
Unconjugated | |
UBE2E2 | |
E2 ubiquitin-conjugating enzyme E2, FLJ25157, UbcH8, ubiquitin carrier protein E2, ubiquitin conjugating enzyme E2E 2, ubiquitin-conjugating enzyme E2E 2, ubiquitin-conjugating enzyme E2E 2 (homologous to yeast UBC4/5), ubiquitin-conjugating enzyme E2E 2 (UBC4/5 homolog, yeast), ubiquitin-protein ligase E2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7325 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q96LR5 | |
UBE2E2 | |
Between 1 and 25 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.