Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2E3 Antibody (OTI1B4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274737
Description
UBE2E3 Monoclonal specifically detects UBE2E3 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
UBE2E3 | |
Monoclonal | |
LYOPH | |
Western Blot 1:500-2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, CyTOF-ready | |
EC 6.3.2.19, UBCE4, UbcH9, UBCH9ubiquitin-conjugating enzyme E2E 3 (homologous to yeast UBC4/5), UbcM2, Ubiquitin carrier protein E3, ubiquitin-conjugating enzyme E2 E3, Ubiquitin-conjugating enzyme E2-23 kDa, ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase E3 | |
Mouse | |
22.7 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF | |
OTI1B4 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
UBE2E3 | |
Full length human recombinant protein of human UBE2E3(NP_006348) produced in E.coli. | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
10477 | |
Human, Mouse, Rat, Canine | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction