Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2E3 Antibody (OTI1B4), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274737PE
Description
UBE2E3 Monoclonal specifically detects UBE2E3 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.Specifications
UBE2E3 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
UBE2E3 | |
Full length recombinant protein of human UBE2E3 (NP_006348) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG2b |
Flow Cytometry | |
OTI1B4 | |
Flow Cytometry | |
EC 6.3.2.19, UBCE4, UbcH9, UBCH9ubiquitin-conjugating enzyme E2E 3 (homologous to yeast UBC4/5), UbcM2, Ubiquitin carrier protein E3, ubiquitin-conjugating enzyme E2 E3, Ubiquitin-conjugating enzyme E2-23 kDa, ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase E3 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
10477.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction