Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2E3 Monoclonal Antibody (OTI1D12), TrueMAB™, OriGene
SDP

Catalog No. 502540183
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0183 30 μL
50-167-5311 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0183 Supplier OriGene Supplier No. TA504671S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBE2E3 Monoclonal antibody specifically detects UBE2E3 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen UBE2E3
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1D12
Concentration 1.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene AHSG
Gene Accession No. P02765
Gene Alias E2 ubiquitin-conjugating enzyme E3; Ubce4; UBCH9; UbcM2; UBE2E3; Ubiquitin carrier protein E3; ubiquitin conjugating enzyme E2 E3; ubiquitin conjugating enzyme E2E 3; ubiquitin-conjugating enzyme 4; ubiquitin-conjugating enzyme E2 E3; Ubiquitin-conjugating enzyme E2-23 kDa; ubiquitin-conjugating enzyme E2E 3; ubiquitin-conjugating enzyme E2E 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2E 3, UBC4/5 homolog; ubiquitin-protein ligase E3
Host Species Mouse
Immunogen Full length human recombinant protein of human AHSG produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.