Learn More
UBE2E3 Mouse anti-Human, Clone: OTI2B1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504672

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein shares 100% sequence identity with the mouse and rat counterparts, which indicates that this enzyme is highly conserved in eukaryotes. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
UBE2E3 | |
Monoclonal | |
Unconjugated | |
UBE2E3 | |
UBCH9, UbcM2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10477 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI2B1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q969T4 | |
UBE2E3 | |
Full length human recombit protein of human UBE2E3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.