Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2E3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA586822
Description
Purity is > 95% by SDS-PAGE.
The ubiquitin (Ub) pathway involves three sequential enzymatic steps that facilitate the conjugation of Ub and Ub-like molecules to specific protein substrates.The first step requires the ATP-dependent activation of the Ub C-terminusand the assembly of multi-Ub chains by the Ub-activating enzyme known as the E1 component. The Ub chain is then conjugated to the Ub-conjugating enzyme (E2) to generate an intermediate Ub-E2 complex. The Ub-ligase (E3) then catalyzes the transfer of Ub from E2 to the appropriate protein substrate. A wide range of enzymes facilitate in the proteolytic Ub pathway including UBE2E3, also designated UBCH9, which catalyzes the covalent attachment of ubiquitin to other proteins and is involved in the regulation of transepithelial sodium transport in renal cells. UBE2E3 may also be involved in cell growth arrest. The UBE2E3 protein shuttles between the cytoplasm and nucleus in a IPO11-dependent manner. It is ubiquitously expressed at low levels and is highly expressed in skeletal muscle.
Specifications
UBE2E3 | |
Polyclonal | |
Unconjugated | |
UBE2E3 | |
E2 ubiquitin-conjugating enzyme E3; Ubce4; UBCH9; UbcM2; UBE2E3; Ubiquitin carrier protein E3; ubiquitin conjugating enzyme E2 E3; ubiquitin conjugating enzyme E2E 3; ubiquitin-conjugating enzyme 4; ubiquitin-conjugating enzyme E2 E3; Ubiquitin-conjugating enzyme E2-23 kDa; ubiquitin-conjugating enzyme E2E 3; ubiquitin-conjugating enzyme E2E 3 (homologous to yeast UBC4/5); ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast); ubiquitin-conjugating enzyme E2E 3, UBC4/5 homolog; ubiquitin-protein ligase E3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10477, 295686 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969T4 | |
UBE2E3 | |
A synthetic peptide corresponding to residues in Human UBE2E3. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction