Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2G1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562212
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562212 Supplier Invitrogen™ Supplier No. PA562212
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RWLPIHTVET IMISVISMLA DPNGDSPANV DAAKEWREDR NGEFKRKVA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2G1 is a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein shares 98-100% sequence identity with the zebrafish, frog, rat and mouse counterparts, which indicates that this enzyme is highly conserved in eukaryotes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2G1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UBE2G1
Gene Accession No. P62253
Gene Alias 2700059C12Rik; AI256795; AU014992; AW552068; D130023C12Rik; E2 ubiquitin-conjugating enzyme G1; E217K; Ubc7; UBE2G; UBE2G1; ubiqu; Ubiquitin carrier protein G1; ubiquitin conjugating enzyme E2 G1; ubiquitin conjugating enzyme E2G 1; ubiquitin-conjugating enzyme E2 G1; Ubiquitin-conjugating enzyme E2 G1, N-terminally processed; ubiquitin-conjugating enzyme E2G 1; ubiquitin-conjugating enzyme E2G 1 (homologous to C. elegans UBC7); ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, C. elegans); ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, yeast); ubiquitin-conjugating enzyme UBC7; ubiquitin-protein ligase G1
Gene Symbols UBE2G1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UBE2G1. Recombinant protein control fragment (Product #RP-100554).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7326
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.