Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2G2 Monoclonal Antibody (OTI4D4), TrueMAB™, OriGene
SDP

Catalog No. 502539830
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9830 30 μL
50-167-5759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9830 Supplier OriGene Supplier No. TA505238S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBE2G2 Monoclonal antibody specifically detects UBE2G2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen UBE2G2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HCFC2
Gene Accession No. Q9Y5Z7
Gene Alias 1110003O05Rik; D10Xrf369; E2 ubiquitin-conjugating enzyme G2; EGK_13178; UBC7; Ubc7p; UBE2G2; Ubiquitin carrier protein G2; ubiquitin conjugating enzyme 7; ubiquitin conjugating enzyme E2 G2; ubiquitin conjugating enzyme E2G 2; ubiquitin conjugating enzyme G2; ubiquitin-conjugating enzyme 7 homolog; ubiquitin-conjugating enzyme E2 G2; ubiquitin-conjugating enzyme E2G 2; ubiquitin-conjugating enzyme E2G 2 (homologous to yeast UBC7); ubiquitin-conjugating enzyme E2G 2 (UBC7 homolog, yeast); ubiquitin-protein ligase G2
Host Species Mouse
Immunogen Full length human recombinant protein of human HCFC2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29915
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.