Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596648 Supplier Invitrogen™ Supplier No. PA596648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSPSPGKRR MDTDVVKLIE SKHEVTILGG LNEFVVKFYG PQGTPYEGGV WKVRVDLPDK YPFKSPSIGF MNKIFHPNID EASGTVCLDV INQTWTALYD LTNIFESFLP QLLAYPNPID PLNGDAAAMY LHRPEEYKQK IKEYIQKYAT EEALKEQEEG TGDSSSESSM SDFSEDEAQD MEL; Positive Samples: SW480, BT-474, 293T, U-251MG, SKOV3, Mouse thymus, Mouse liver, Mouse lung, Rat lung.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein sequence is 100% identical to the mouse homolog and 98% identical to the frog and zebrafish homologs. Three alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2H
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE2H
Gene Accession No. P62256, P62257
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme H; 1500009C23Rik; AI181839; AI326965; AI462483; AW227540; E2 ubiquitin-conjugating enzyme H; E2-20K; fc07a05; fi24c01; GID complex subunit 3, UBC8 homolog; Gid3; N28148; si:bz3c13.2; Ubc8; UBCH; UbcH2; ube2h; Ubiquitin carrier protein H; ubiquitin conjugating enzyme E2 H; ubiquitin conjugating enzyme E2H; ubiquitin-conjugating enzyme E2 H; Ubiquitin-conjugating enzyme E2-20K; ubiquitin-conjugating enzyme E2H; ubiquitin-conjugating enzyme E2H (homologous to yeast UBC8); ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast); ubiquitin-protein ligase H; Unknown (protein for MGC:134106); wu:fc07a05; wu:fi24c01
Gene Symbols UBE2H
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-183 of human UBE2H (NP_0033351).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22214, 296956, 7328
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.