Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ube2H Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988504
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8504 Supplier Bioss Supplier No. BS8370R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein sequence is 100% identical to the mouse homolog and 98% identical to the frog and zebrafish homologs. Three alternatively spliced transcript variants have been found for this gene and they encode distinct isoforms.

Specifications

Antigen Ube2H
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UBE2H
Gene Accession No. P62256
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme H; 1500009C23Rik; AI181839; AI326965; AI462483; AW227540; BOS_4477; E2 ubiquitin-conjugating enzyme H; E2-20K; fc07a05; fi24c01; GID complex subunit 3, UBC8 homolog; GID3; LOC100350643; N28148; si:bz3c13.2; UBC8; UBCH; UbcH2; UBE2H; Ubiquitin carrier protein H; ubiquitin conjugating enzyme E2 H; ubiquitin conjugating enzyme E2H; Ubiquitin-conjugating enzyme E2 H; Ubiquitin-conjugating enzyme E2-20K; ubiquitin-conjugating enzyme E2H; ubiquitin-conjugating enzyme E2H (homologous to yeast UBC8); ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast); ubiquitin-conjugating enzyme-like; Ubiquitin-protein ligase H; wu:fc07a05; wu:fi24c01
Gene Symbols UBE2H
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Ube2H/UBCH2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7328
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.