Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2J1 Antibody (OTI3D1), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274747MFV500
Description
UBE2J1 Monoclonal antibody specifically detects UBE2J1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow CytometrySpecifications
UBE2J1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
51465 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
OTI3D1 | |
Flow Cytometry | |
CGI-76, EC 6.3.2.19, HSPC153, HSPC205, HSU93243, HsUBC6e, MGC12555, NCUBE-1, NCUBE1HSUBC6e, Non-canonical ubiquitin-conjugating enzyme 1, non-canonical ubquitin conjugating enzyme 1, Ubc6p, ubiquitin-conjugating enzyme E2 J1, ubiquitin-conjugating enzyme E2, J1 (UBC6 homolog, yeast), Yeast ubiquitin-conjugating enzyme UBC6 homolog E | |
Full length human recombinant protein of human UBE2J1(NP_057105) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction