Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2J1 Monoclonal Antibody (OTI2G2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505035S


Description
UBE2J1 Monoclonal antibody specifically detects UBE2J1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
UBE2J1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P35250 | |
Mouse | |
Affinity chromatography | |
RUO | |
5982 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Flow Cytometry, Western Blot | |
OTI2G2 | |
Unconjugated | |
RFC2 | |
0710008M05Rik; 1110030I22Rik; CGI-76; E2 ubiquitin-conjugating enzyme J1; HSPC153; HSPC205; HSU93243; HsUBC6e; MGC12555; Ncube; NCUBE1; NCUBE-1; non-canonical ubiquitin conjugating enzyme 1; non-canonical ubiquitin-conjugating enzyme 1; UBC6; UBC6E; Ubc6p; Ube2j1; ubiquitin conjugating enzyme E2 J1; ubiquitin conjugating enzyme E2, J1; ubiquitin-conjugating enzyme E2 J1; ubiquitin-conjugating enzyme E2, J1; ubiquitin-conjugating enzyme E2, J1 (UBC6 homolog, yeast); ubiquitin-conjugating enzyme E2, J1, U; ubiquitin-conjugating enzyme E2J 1; yeast ubiquitin-conjugating enzyme UBC6 homolog E; zgc:63554 | |
Human recombinant protein fragment corresponding to amino acids 1-234 of human RFC2 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction