Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2J1 Monoclonal Antibody (OTI3D5), TrueMAB™, OriGene
SDP

Catalog No. 502539979
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9979 30 μL
50-167-5566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9979 Supplier OriGene Supplier No. TA505006S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBE2J1 Monoclonal antibody specifically detects UBE2J1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen UBE2J1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ftcd
Gene Accession No. O95954
Gene Alias 0710008M05Rik; 1110030I22Rik; CGI-76; E2 ubiquitin-conjugating enzyme J1; HSPC153; HSPC205; HSU93243; HsUBC6e; MGC12555; Ncube; NCUBE1; NCUBE-1; non-canonical ubiquitin conjugating enzyme 1; non-canonical ubiquitin-conjugating enzyme 1; UBC6; UBC6E; Ubc6p; Ube2j1; ubiquitin conjugating enzyme E2 J1; ubiquitin conjugating enzyme E2, J1; ubiquitin-conjugating enzyme E2 J1; ubiquitin-conjugating enzyme E2, J1; ubiquitin-conjugating enzyme E2, J1 (UBC6 homolog, yeast); ubiquitin-conjugating enzyme E2, J1, U; ubiquitin-conjugating enzyme E2J 1; yeast ubiquitin-conjugating enzyme UBC6 homolog E; zgc:63554
Host Species Mouse
Immunogen Full length human recombinant protein of human FTCD produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10841
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.