Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2J1 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991904
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1904 100 μg
50-199-1905 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1904 Supplier Rockland Immunochemicals Supplier No. 600401991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against human Ube2j1 protein. A BLAST analysis was used to suggest cross-reactivity with Ube2j1 protein from human, mouse and rat based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum and may contribute to quality control ER-associated degradation by the ubiquitin-proteasome system.

Specifications

Antigen UBE2J1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Ube2j1
Gene Accession No. Q9Y385
Gene Alias 0710008M05Rik; 1110030I22Rik; CGI-76; E2 ubiquitin-conjugating enzyme J1; HSPC153; HSPC205; HSU93243; HsUBC6e; LOC100341530; MGC12555; Ncube; Ncube1; NCUBE-1; non-canonical ubiquitin conjugating enzyme 1; Non-canonical ubiquitin-conjugating enzyme 1; UBC6; UBC6E; Ubc6p; Ube2j1; ubiquitin conjugating enzyme E2 J1; ubiquitin conjugating enzyme E2, J1; Ubiquitin-conjugating enzyme E2 J1; ubiquitin-conjugating enzyme E2, J1; ubiquitin-conjugating enzyme E2, J1 (UBC6 homolog, yeast); ubiquitin-conjugating enzyme E2, J1, U; ubiquitin-conjugating enzyme E2J 1; Yeast ubiquitin-conjugating enzyme UBC6 homolog E; zgc:63554
Gene Symbols Ube2j1
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51465
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.