Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2J2 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501992001
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2001 25 μL
50-199-2000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-2001 Supplier Rockland Immunochemicals Supplier No. 600401B43S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against human Ube2j2 protein. A BLAST analysis was used to suggest cross-reactivity with Ube2j2 protein from human, horse, opossum, cattle, dog, mouse, rat, macaque, salmon, chicken, zebra finch, Xenopus and platypus based on 100% homology with the immunizing sequence. Reactivity against homologues from other sources is not known.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2J2 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is located in the membrane of the endoplasmic reticulum. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined.

Specifications

Antigen UBE2J2
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene UBE2J2
Gene Accession No. Q6P073, Q8N2K1
Gene Alias 1200007B18Rik; 2400008G19Rik; 5730472G04Rik; AL022923; BOS_15873; E2 ubiquitin-conjugating enzyme J2; LOC100350316; NCUBE2; NCUBE-2; Non-canonical ubiquitin-conjugating enzyme 2; PRO2121; Ubc6; Ubc6p; Ubc6p homolog; UBE2J2; ubiquitin conjugating enzyme 6; ubiquitin conjugating enzyme E2 J2; ubiquitin conjugating enzyme E2, J2; Ubiquitin-conjugating enzyme E2 J2; ubiquitin-conjugating enzyme E2, J2; ubiquitin-conjugating enzyme E2, J2 (UBC6 homolog, yeast); ubiquitin-conjugating enzyme E2, J2 homolog; ubiquitin-conjugating enzyme E2J 2; yeast UBC6 homolog; zgc:162164
Gene Symbols UBE2J2
Host Species Rabbit
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118424, 140499, 298689
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.