Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2J2/UBC6 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306347JF646
Description
UBE2J2/UBC6 Polyclonal antibody specifically detects UBE2J2/UBC6 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
UBE2J2/UBC6 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
EC 6.3.2.19, J2, Ubc6p, ubiquitin-conjugating enzyme E2 J2, ubiquitin-conjugating enzyme E2, J2 (UBC6 homolog, yeast), yeast UBC6 homolog | |
Produced in rabbits immunized with E. coli-derived Human UBE2J2/UBC6 fragment. | |
0.1 mL | |
Stem Cell Markers | |
118424 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction