Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2K/E2-25K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Applications | Western Blot |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Host Species | Rabbit |
Description
UBE2K/E2-25K Polyclonal specifically detects UBE2K/E2-25K in Human samples. It is validated for Western Blot.Specifications
Western Blot | |
Unconjugated | |
Rabbit | |
P61086 | |
UBE2K | |
Primary | |
22 kDa |
Polyclonal | |
Purified | |
RUO | |
3093 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title