Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2K/E2-25K Sheep anti-Human, Mouse, Rat, Polyclonal, R&D Systems™

Catalog No. AF6609SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6609SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6609SP Supplier R&D Systems Supplier No. AF6609SP
This item is not returnable. View return policy

Sheep Polyclonal Antibody

UBE2K/E2-25K Polyclonal antibody AF6609SP specifically detects UBE2K/E2-25K in Human, Mouse, Rat samples. It is validated for Western Blotting

Specifications

Antigen UBE2K/E2-25K
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P61086
Gene Alias E2(25K), E225K, E2-25K, EC 6.3.2.19, Hip2, HIP-2, huntingtin interacting protein 2, Huntingtin-interacting protein 2, HYPG, LIG, UbcH1, UBE2K, UBE2K-25K, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 K, Ubiquitin-conjugating enzyme E2(25K), Ubiquitin-conjugating enzyme E2-25 kDa, Ubiquitin-conjugating enzyme E2-25K, ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast), ubiquitin-protein ligase
Gene Symbols UBE2K
Host Species Sheep
Immunogen E. coli-derived recombinant human UBE2K/E2-25K, Ala2-Asn200, Accession # P61086
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3093
Test Specificity Detects human, mouse, and rat UBE2K/E2-25K in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.