Learn More
Invitrogen™ UBE2L3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592154
Description
Immunogen sequence: RKCGMKNFRN IQVDEANLLT WQGLIVPDNP PYDKGAFRIE INFPAEYPFK PPKITFKTKI YHPNIDEKGQ VCLPVISAEN WKPATKTDQV IQSLIALVND PQPEHPLRAD LAEEYSKDRK KFCKNAEEFT KKYGEKRPVD; Positive Samples: U-251MG, HL-60, K-562, HeLa, Mouse testis, Mouse brain, Mouse skeletal muscle; Cellular Location: Cytoplasm, Nucleus.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L3 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to participate in the ubiquitination of p53, c-Fos, and the NF-kB precursor p105 in vitro.
Specifications
UBE2L3 | |
Polyclonal | |
Unconjugated | |
UBE2L3 | |
C79827; E2 ubiquitin-conjugating enzyme L3; E2-F1; L-UBC; UBCE7; UBCH7; UbcM4; Ube2l3; Ubiquitin carrier protein L3; ubiquitin conjugating enzyme E2 L3; ubiquitin conjugating enzyme E2L 3; ubiquitin-conjugating enzyme 7; ubiquitin-conjugating enzyme E2 L3; Ubiquitin-conjugating enzyme E2-F1; ubiquitin-conjugating enzyme E2L 3; ubiquitin-conjugating enzyme UBCH7; ubiquitin-protein ligase L3; Unknown (protein for MGC:127956) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
22195, 363836, 7332 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P68036, P68037 | |
UBE2L3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 73-212 of human UBE2L3 (NP_0012432841). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.