Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2L6 Recombinant Rabbit Monoclonal Antibody (140)
GREENER_CHOICE

Catalog No. PIMA530984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530984 Supplier Invitrogen™ Supplier No. MA530984
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human UBCH8/UBE2L6.

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L6 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to function in the E6/E6-ap-induced ubiquitination of p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2L6
Applications ELISA
Classification Recombinant Monoclonal
Clone 140
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene UBE2L6
Gene Accession No. O14933
Gene Alias 2810489I21Rik; E2 ubiquitin conjugating enzyme; E2 ubiquitin-conjugating enzyme L6; MGC40331; retinoic acid induced gene B protein; retinoic acid-induced gene B protein; RIG-B; Ubce8; UBCH8; Ubcm8; UBE2L6; Ubiquitin carrier protein L6; ubiquitin conjugating enzyme E2 L6; ubiquitin conjugating enzyme E2L 6; Ubiquitin/ISG15-conjugating enzyme E2 L6; ubiquitin-conjugating enzyme 8; ubiquitin-conjugating enzyme E2L 6; Ubiquitin-protein ligase L6
Gene Symbols UBE2L6
Host Species Rabbit
Immunogen Recombinant Human UBCH8/UBE2L6 protein (Met1-Ser153).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9246
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.