Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2M Monoclonal Antibody (OTI2D9)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525739

Catalog No. PIMA525739


Only null left
Add to Cart

Description

Description

UBE2M Monoclonal Antibody for Western Blot, ICC/IF

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is linked with a ubiquitin-like protein, NEDD8, which can be conjugated to cellular proteins, such as Cdc53/culin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

UBE2M
Monoclonal
0.69 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P61081, P61082
UBE2M
Full length human recombinant protein of UBE2M produced in HEK293T cell.
100 μL
Primary
Human, Mouse, Canine, Monkey
Antibody
IgG1
Western Blot, Immunocytochemistry
OTI2D9
Unconjugated
UBE2M
2510040H03Rik; EGK_11185; hUbc12; NEDD8 carrier protein; NEDD8 protein ligase; NEDD8-conjugating enzyme Ubc12; UBC12; UBC12 homolog, yeast; UBC-RS2; Ube2m; ubiquitin C, related sequence 2; ubiquitin carrier protein M; ubiquitin conjugating enzyme E2 M; ubiquitin conjugating enzyme E2M; Ubiquitin-conjugating enzyme E2 M; ubiquitin-conjugating enzyme E2M; ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast); ubiquitin-protein ligase M; yeast UBC12 homolog
Mouse
Affinity chromatography
RUO
22192, 484221, 9040
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.