Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2M Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542283
Description
Peptide sequence: IKLFSLKQQK KEEESAGGTK GSSKKASAAQ LRIQKDINEL NLPKTCDISF Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is linked with a ubiquitin-like protein, NEDD8, which can be conjugated to cellular proteins, such as Cdc53/culin.
Specifications
UBE2M | |
Polyclonal | |
Unconjugated | |
UBE2M | |
2510040H03Rik; EGK_11185; hUbc12; NEDD8 carrier protein; NEDD8 protein ligase; NEDD8-conjugating enzyme Ubc12; UBC12; UBC12 homolog, yeast; UBC-RS2; Ube2m; ubiquitin C, related sequence 2; ubiquitin carrier protein M; ubiquitin conjugating enzyme E2 M; ubiquitin conjugating enzyme E2M; Ubiquitin-conjugating enzyme E2 M; ubiquitin-conjugating enzyme E2M; ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast); ubiquitin-protein ligase M; yeast UBC12 homolog | |
Rabbit | |
Affinity chromatography | |
RUO | |
22192, 9040 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P61081, P61082 | |
UBE2M | |
Synthetic peptide directed towards the N-terminal of human UBE2M (aa 2-51). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction