Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2M/Ubc12 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298834F
Description
UBE2M/Ubc12 Polyclonal antibody specifically detects UBE2M/Ubc12 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
UBE2M/Ubc12 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human UBE2M/Ubc12. | |
0.1 mL | |
Biologically Active Proteins | |
9040 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction