Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2M/Ubc12 Antibody (OTI2D9), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274748AF700
Description
UBE2M/Ubc12 Monoclonal specifically detects UBE2M/Ubc12 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
UBE2M/Ubc12 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
UBE2M | |
Full length recombinant protein of human UBE2M/Ubc12 (NP_003960) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey, Primate | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2D9 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins | |
9040.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction