Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ube2N Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988488
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8488 Supplier Bioss Supplier No. BS8351R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Ubc13 is a monoclonal antibody specific to the ˜19 kDa Ubc13 protein. Ubc13 is a ubiquitin conjugating enzyme that is complexed with a ubiquitin activating enzyme (E1), Mms2. The association of Mms2 to Ubc13 forms an active heterodimer that is involved in error-free post replicative DNA repair in yeast. Upon complexing with another conjugating enzyme variant, Uev1a, Ubc13 participates in the regulation of NF-kB signal transduction pathway in humans.

Specifications

Antigen Ube2N
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UBE2N
Gene Accession No. P61088
Gene Alias 1500026J17Rik; AL022654; BB101821; bendless protein; bendless-like ubiquitin conjugating enzyme; Bendless-like ubiquitin-conjugating enzyme; BLU; E2 ubiquitin-conjugating enzyme N; epididymis secretory protein Li 71; HEL-S-71; MGC131857; MGC8489; Ubc13; UbcH13; UbcH-ben; UBCHBEN; UBC13; Ube2n; Ubiquitin carrier protein N; ubiquitin conjugating enzyme E2 N; ubiquitin conjugating enzyme E2N; Ubiquitin-conjugating enzyme E2 N; ubiquitin-conjugating enzyme E2N; ubiquitin-conjugating enzyme E2N (homologous to yeast UBC13); ubiquitin-conjugating enzyme E2N (UBC13 homolog, yeast); Ubiquitin-protein ligase N; yeast UBC13 homolog
Gene Symbols UBE2N
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Ube2N/UBC13.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7334
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.