Learn More
UBE2NL Mouse anti-Human, Clone: OTI8E6, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF812448

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is intronless and encodes a member of the ubiquitin-conjugating enzyme family. The protein product is 91% identical to ubiquitin-conjugating enzyme E2N, a multi-exon gene product. This locus represents a polymorphic pseudogene, where some individuals contain an allele that can encode a full-length protein, while others have a non-functional allele containing a premature stop codon (reference SNP rs237520) that truncates the coding sequence.Specifications
UBE2NL | |
Monoclonal | |
Unconjugated | |
UBE2NL | |
Li174 | |
Mouse | |
Affinity Chromatography | |
RUO | |
389898 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8E6 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q5JXB2 | |
UBE2NL | |
Human recombit protein fragment corresponding to amino acids 1-100 of human UBE2NL produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.