Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2NL Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548403
Description
Predicted to react with bovine, monkey, mouse and rat based on sequence homology.
This gene is intronless and encodes a member of the ubiquitin-conjugating enzyme family. The protein product is 91% identical to ubiquitin-conjugating enzyme E2N, a multi-exon gene product. This locus represents a polymorphic pseudogene, where some individuals contain an allele that can encode a full-length protein, while others have a non-functional allele containing a premature stop codon (reference SNP rs237520) that truncates the coding sequence. [provided by RefSeq, Jun 2014]
Specifications
UBE2NL | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q5JXB2 | |
UBE2NL | |
KLH conjugated synthetic peptide between 6-34 amino acids from the N-terminal region of human UBE2NL. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
UBE2NL | |
Putative ubiquitin-conjugating enzyme E2 N-like, Epididymis tissue protein Li 174, UBE2NL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
389898 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction