Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2O Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5142032
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000. Preliminary analysis showed no signal in Western Blot.
E2/E3 hybrid ubiquitin-protein ligase that displays both E2 and E3 ligase activities and mediates monoubiquitination of target proteins (PubMed:23455153, PubMed:24703950). Negatively regulates TRAF6-mediated NF-kappa-B activation independently of its E2 activity (PubMed:23381138). Acts as a positive regulator of BMP7 signaling by mediating monoubiquitination of SMAD6, thereby regulating adipogenesis (PubMed:23455153). Mediates monoubiquitination at different sites of the nuclear localization signal (NLS) of BAP1, leading to cytoplasmic retention of BAP1. Also able to monoubiquitinate the NLS of other chromatin-associated proteins, such as INO80 and CXXC1, affecting their subcellular location (PubMed:24703950). Acts as a regulator of retrograde transport by assisting the TRIM27:MAGEL2 E3 ubiquitin ligase complex to mediate 'Lys-63'-linked ubiquitination of WASHC1, leading to promote endosomal F-actin assembly (PubMed:23452853). [UniProt].
Specifications
UBE2O | |
Polyclonal | |
Unconjugated | |
UBE2O | |
(E3-independent) E2 ubiquitin-conjugating enzyme; (E3-independent) E2 ubiquitin-conjugating enzyme UBE2O; 9630022H21; B230113M03Rik; E2/E3 hybrid ubiquitin-protein ligase UBE2O; E2-230K; Kiaa1734; mKIAA1734; RGD1310297; UBE2O; Ubiquitin carrier protein O; ubiquitin conjugating enzyme E2 O; ubiquitin conjugating enzyme E2O; ubiquitinating enzyme E2-230 kDa; ubiquitin-conjugating enzyme E2 O; Ubiquitin-conjugating enzyme E2 of 230 kDa; ubiquitin-conjugating enzyme E2-230K; ubiquitin-conjugating enzyme E2O; ubiquitin-protein ligase O | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
63893 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9C0C9 | |
UBE2O | |
Peptide with sequence ALLEAGMPECTEDK. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction