Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2O Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598873
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598873 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598873 Supplier Thermo Scientific Supplier No. PA598873
Add to Cart
Add to Cart

UBE2O Polyclonal Antibody

UBE2O Polyclonal Antibody for Western Blot, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2O
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene UBE2O
Gene Accession No. Q9C0C9
Gene Alias (E3-independent) E2 ubiquitin-conjugating enzyme; (E3-independent) E2 ubiquitin-conjugating enzyme UBE2O; 9630022H21; B230113M03Rik; E2/E3 hybrid ubiquitin-protein ligase UBE2O; E2-230K; Kiaa1734; mKIAA1734; RGD1310297; UBE2O; Ubiquitin carrier protein O; ubiquitin conjugating enzyme E2 O; ubiquitin conjugating enzyme E2O; ubiquitinating enzyme E2-230 kDa; ubiquitin-conjugating enzyme E2 O; Ubiquitin-conjugating enzyme E2 of 230 kDa; ubiquitin-conjugating enzyme E2-230K; ubiquitin-conjugating enzyme E2O; ubiquitin-protein ligase O
Gene Symbols UBE2O
Host Species Rabbit
Immunogen Recombinant Human (E3-independent) E2 ubiquitin-conjugating enzyme protein (10-277AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63893
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.