Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2O Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30321620UL
Description
UBE2O Polyclonal specifically detects UBE2O in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Knockout Validated.Specifications
UBE2O | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
63893 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence, Immunoprecipitation, KnockDown | |
Unconjugated | |
E2-230K, EC 6.3.2.19, FLJ12878, KIAA1734, Ubiquitin carrier protein O, ubiquitin-conjugating enzyme E2 O, Ubiquitin-conjugating enzyme E2 of 230 kDa, Ubiquitin-conjugating enzyme E2-230K, ubiquitin-conjugating enzyme E2O, Ubiquitin-protein ligase O | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1073-1292 of human UBE2O (NP_071349.3). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction