Learn More
Description
Specifications
Specifications
| Antigen | UBE2Q1 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulation | PBS (pH 7.2), 40% Glycerol |
| Gene Alias | EC 6.3.2.19, galactosyl transferase-associated protein, GTAP, NICE5, NICE-5, PRO3094, Protein NICE-5, UBE2Q, Ubiquitin carrier protein Q1, ubiquitin-conjugating enzyme E2 Q1, ubiquitin-conjugating enzyme E2Q (putative), ubiquitin-conjugating enzyme E2Q (putative) 1, ubiquitin-conjugating enzyme E2Q family member 1, Ubiquitin-protein ligase Q1 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: PCLRRELKLLESIFHRGHERFRIASACLDELSCEFLLAG |
| Purification Method | Immunogen affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
