Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2Q1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS8383R

Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin-protein ligases (E3s). This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein is 98% identical to the mouse counterpart.Specifications
UBE2Q1 | |
Polyclonal | |
Unconjugated | |
UBE2Q1 | |
1110002C01Rik; 2310012M18Rik; AI854014; BOS_2868; E2 ubiquitin-conjugating enzyme Q1; Galactosyl transferase-associated protein; GTAP; hypothetical protein LOC553603; LOC100347500; NICE5; NICE-5; PRO3094; Protein NICE-5; Ube2q; Ube2q1; Ubiquitin carrier protein Q1; ubiquitin conjugating enzyme E2 Q1; ubiquitin conjugating enzyme E2Q family member 1; Ubiquitin-conjugating enzyme E2 Q1; ubiquitin-conjugating enzyme E2 Q2; ubiquitin-conjugating enzyme E2Q; ubiquitin-conjugating enzyme E2Q (putative) 1; ubiquitin-conjugating enzyme E2Q family member 1; Ubiquitin-protein ligase Q1; wu:fi42b06; zgc:110254 | |
Rabbit | |
Protein A | |
RUO | |
55585 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q7Z7E8 | |
UBE2Q1 | |
KLH conjugated synthetic peptide derived from human UBE2Q1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction