Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2Q2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580202
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, A431 whole cell, MCF-7 whole cell, SW620 whole cell. IHC: human lung cancer tissue. ICC/IF: U20S cell. Flow: A549 cell.
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes 'Lys-48'-linked polyubiquitination. [UniProt].
Specifications
UBE2Q2 | |
Polyclonal | |
Unconjugated | |
UBE2Q2 | |
3010021M21Rik; DKFZp762C143; E2 ubiquitin-conjugating enzyme Q2; RGD1307680; UBE2Q2; Ubiquitin carrier protein Q2; ubiquitin conjugating enzyme E2 Q2; ubiquitin conjugating enzyme E2Q family member 2; ubiquitin conjugating enzyme-implantation induced; ubiquitin-conjugating enzyme E2 Q2; ubiquitin-conjugating enzyme E2Q (putative) 2; ubiquitin-conjugating enzyme E2Q 2; ubiquitin-conjugating enzyme E2Q family member 2; ubiquitin-conjugating enzyme UBCi; Ubiquitin-protein ligase Q2; Unknown (protein for MGC:134315); wu:fi34a03; zgc:56219 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92912 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
Q8WVN8 | |
UBE2Q2 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human UBE2Q2 (83-123aa LERLEDTKNNNLLRQQLKWLICELCSLYNLPKHLDVEMLDQ). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction