Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2Q2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597324 Supplier Invitrogen™ Supplier No. PA597324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSVSGLKAEL KFLASIFDKN HERFRIVSWK LDELHCQFLV PQQGSPHSLP PPLTLHCNIT ESYPSSSPIW FVDSEDPNLT SVLERLEDTK NNNLLRQQLK WLICELCSLY NLPKHLDVEM LDQPLPTGQN GTTEEVTSEE EEEEEEMAED; Positive Samples: SKOV3, HeLa, HL-60, A-549; Cellular Location: Cytoplasm.

Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro catalyzes 'Lys-48'-linked polyubiquitination. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2Q2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE2Q2
Gene Accession No. Q8K2Z8, Q8WVN8
Gene Alias 3010021M21Rik; DKFZp762C143; E2 ubiquitin-conjugating enzyme Q2; RGD1307680; UBE2Q2; Ubiquitin carrier protein Q2; ubiquitin conjugating enzyme E2 Q2; ubiquitin conjugating enzyme E2Q family member 2; ubiquitin conjugating enzyme-implantation induced; ubiquitin-conjugating enzyme E2 Q2; ubiquitin-conjugating enzyme E2Q (putative) 2; ubiquitin-conjugating enzyme E2Q 2; ubiquitin-conjugating enzyme E2Q family member 2; ubiquitin-conjugating enzyme UBCi; Ubiquitin-protein ligase Q2; Unknown (protein for MGC:134315); wu:fi34a03; zgc:56219
Gene Symbols UBE2Q2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109161, 92912
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.