Learn More
UBE2Q2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | UBE2Q2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-3908
![]() |
Bethyl Laboratories, Inc
A302961AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Rabbit
Catalyzes the covalent attachment of ubiquitin to other proteins (By similarity).Specifications
UBE2Q2 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
DKFZp762C143, E2 ubiquitin-conjugating enzyme Q2, ubiquitin carrier protein Q2, ubiquitin conjugating enzyme E2Q family member 2, ubiquitin-conjugating enzyme E2Q (putative) 2, ubiquitin-conjugating enzyme E2Q family member 2, ubiquitin-protein ligase Q2 | |
Between 325 and 375 | |
Primary | |
4° C | |
UBE2Q2 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8K2Z8, Q8WVN8 | |
109161, 92912 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.