Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2Q2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501563906
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-3906 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-3906 Supplier Bethyl Laboratories, Inc Supplier No. A302961A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Rabbit

Catalyzes the covalent attachment of ubiquitin to other proteins (By similarity).

Specifications

Antigen UBE2Q2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene UBE2Q2
Gene Accession No. Q8K2Z8, Q8WVN8
Gene Alias DKFZp762C143, E2 ubiquitin-conjugating enzyme Q2, ubiquitin carrier protein Q2, ubiquitin conjugating enzyme E2Q family member 2, ubiquitin-conjugating enzyme E2Q (putative) 2, ubiquitin-conjugating enzyme E2Q family member 2, ubiquitin-protein ligase Q2
Gene Symbols UBE2Q2
Host Species Rabbit
Immunogen Between 325 and 375
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109161, 92912
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.