Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2R1/CDC34 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306560MFV450
Description
UBE2R1/CDC34 Polyclonal antibody specifically detects UBE2R1/CDC34 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
UBE2R1/CDC34 | |
Polyclonal | |
Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
cell division cycle 34, cell division cycle 34 homolog (S. cerevisiae), E2-CDC34, EC 6.3.2.19, UBC3, UBE2R1UBCH3, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 R1, Ubiquitin-conjugating enzyme E2-32 kDa complementing, Ubiquitin-conjugating enzyme E2-CDC34, Ubiquitin-protein ligase R1 | |
Produced in rabbits immunized with E. coli-derived Human UBE2R1/CDC34 fragment. | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
997 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction