Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2S Antibody (OTI1B5), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274749V
Description
UBE2S Monoclonal specifically detects UBE2S in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
UBE2S | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
UBE2S | |
Full length recombinant protein of human UBE2S (NP_055316) produced in E.coli. | |
0.1 mL | |
27338.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
OTI1B5 | |
Western Blot, Flow Cytometry | |
E2-EPF5, E2-EPFEC 6.3.2.19, E2EPFEPF5, Ubiquitin carrier protein S, ubiquitin-conjugating enzyme E2 S, ubiquitin-conjugating enzyme E2-24 kD, Ubiquitin-conjugating enzyme E2-24 kDa, Ubiquitin-conjugating enzyme E2-EPF5, ubiquitin-conjugating enzyme E2S, Ubiquitin-protein ligase S | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction