Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2S Antibody (OTI1B5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246279
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry
UBE2S Monoclonal specifically detects UBE2S in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
UBE2S | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Flow Cytometry 1:100 | |
E2-EPF5, E2-EPFEC 6.3.2.19, E2EPFEPF5, Ubiquitin carrier protein S, ubiquitin-conjugating enzyme E2 S, ubiquitin-conjugating enzyme E2-24 kD, Ubiquitin-conjugating enzyme E2-24 kDa, Ubiquitin-conjugating enzyme E2-EPF5, ubiquitin-conjugating enzyme E2S, Ubiquitin-protein ligase S | |
Mouse | |
23.7 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry | |
OTI1B5 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
UBE2S | |
Full length human recombinant protein of human UBE2S(NP_055316) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
27338 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction