Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2S Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530529
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530529 Supplier Invitrogen™ Supplier No. PA530529
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HepG2, HCT116. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (81%), Xenopus laevis (99%), Dog (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the ubiquitin-conjugating enzyme family. The encoded protein is able to form a thiol ester linkage with ubiquitin in a ubiquitin activating enzyme-dependent manner, a characteristic property of ubiquitin carrier proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2S
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene UBE2S
Gene Accession No. Q16763
Gene Alias 0910001J09Rik; 6720465F12Rik; AA409170; E2 ubiquitin-conjugating enzyme S; E2EPF; E2-EPF; E2-EPF5; EPF5; OK/SW-cl.73; RGD1564746; UBE2S; Ubiquitin carrier protein S; ubiquitin conjugating enzyme E2 S; ubiquitin conjugating enzyme E2S; ubiquitin-conjugating enzyme E2 S; ubiquitin-conjugating enzyme E2-24 kD; ubiquitin-conjugating enzyme E2-24 kDa; Ubiquitin-conjugating enzyme E2-EPF5; ubiquitin-conjugating enzyme E2S; ubiquitin-protein ligase S
Gene Symbols UBE2S
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human UBE2S. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27338
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.