Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE2T Antibody (OTI2F5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP202965
Description
UBE2T Monoclonal specifically detects UBE2T in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
UBE2T | |
Monoclonal | |
0.33 mg/mL | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:100 | |
UBE2T | |
Full length human recombinant protein of human UBE2T (NP_054895) produced in HEK293T cell. | |
Affinity Purified | |
RUO | |
29089 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI2F5 | |
Unconjugated | |
Cell proliferation-inducing gene 50 protein, EC 6.3.2.19, HSPC150, HSPC150 protein similar to ubiquitin-conjugating enzyme, PIG50, Ubiquitin carrier protein T, ubiquitin conjugating enzyme, ubiquitin-conjugating enzyme E2 T, ubiquitin-conjugating enzyme E2T (putative), Ubiquitin-protein ligase T | |
Mouse | |
22.3 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction