Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2T Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593247 Supplier Thermo Scientific Supplier No. PA593247
Add to Cart
Add to Cart

UBE2T Polyclonal Antibody

UBE2T Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2T
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE2T
Gene Accession No. Q9CQ37, Q9NPD8
Gene Alias 2700084L22Rik; C80607; Cell proliferation-inducing gene 50 protein; E2 ubiquitin-conjugating enzyme T; FANCT; HSPC150; HSPC150 protein similar to ubiquitin-conjugating enzyme; PIG50; RGD1310816; Ube2t; Ubiquitin carrier protein T; ubiquitin conjugating enzyme E2 T; ubiquitin conjugating enzyme E2T; ubiquitin-conjugating enzyme E2 T; ubiquitin-conjugating enzyme E2T; ubiquitin-conjugating enzyme E2T (putative); ubiquitin-protein ligase T
Gene Symbols UBE2T
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-197 of human UBE2T (NP_0548951).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29089, 360847, 67196
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.