Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2W Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988518
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8518 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8518 Supplier Bioss Supplier No. BS8385R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2W accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins, and catalyzes monoubiquitination. Further, UBE2W is involved in degradation of misfolded chaperone substrates by mediating monoubiquitination of STUB1/CHIP, leading to recruitment of ATXN3 to monoubiquitinated STUB1/CHIP, and restriction of the length of ubiquitin chain attached to STUB1/CHIP substrates by ATXN3. After UV irradiation, but not after mitomycin-C (MMC) treatment, acts as a specific E2 ubiquitin-conjugating enzyme for the Fanconi anemia complex by associating with E3 ubiquitin-protein ligase FANCL and catalyzing monoubiquitination of FANCD2, a key step in the DNA damage pathway. UBE2W In vitro catalyzes ′Lys-11′-linked polyubiquitination, and transfers ubiquitin in complex with RING/U-box type E3s that do not have active site cysteine residues to form thioester bonds with ubiquitin, and preferentially ubiquitinates the N-terminus of substrates, such as ATXN3, STUB1 and SUMO2.

Specifications

Antigen UBE2W
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UBE2W
Gene Accession No. Q96B02
Gene Alias 6130401J04Rik; E2 ubiquitin-conjugating enzyme W; N-terminal E2 ubiquitin-conjugating enzyme; N-terminus-conjugating E2; probable ubiquitin-conjugating enzyme E2 W; UBC16; UBC-16; Ube2w; Ubiquitin carrier protein W; ubiquitin conjugating enzyme E2 W (putative); ubiquitin conjugating enzyme E2W (putative); Ubiquitin-conjugating enzyme 16; ubiquitin-conjugating enzyme 2W; Ubiquitin-conjugating enzyme E2 W; ubiquitin-conjugating enzyme E2 W-like; ubiquitin-conjugating enzyme E2W; ubiquitin-conjugating enzyme E2W (putative); Ubiquitin-protein ligase W
Gene Symbols UBE2W
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human UBE2W.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55284
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.