Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UBE2Z Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577175
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
Specifications
UBE2Z | |
Polyclonal | |
Unconjugated | |
UBE2Z | |
AW049199; C030047H17Rik; D11Moh35; E2 ubiquitin-conjugating enzyme Z; HOYS7; RGD1308347; uba6-specific E2 conjugating enzyme 1; UBA6-specific enzyme E2; UBE2Z; Ubiquitin carrier protein Z; ubiquitin conjugating enzyme E2 Z; ubiquitin conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2 Z; ubiquitin-conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2Z (putative); ubiquitin-protein ligase Z; Use1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
268470, 65264 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3UE37, Q9H832 | |
UBE2Z | |
Recombinant full length Human UBE2Z. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction