Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE2Z Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598930
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598930 Supplier Invitrogen™ Supplier No. PA598930
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2Z Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes an enzyme which ubiquitinates proteins which participate in signaling pathways and apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE2Z
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE2Z
Gene Accession No. Q9H832
Gene Alias AW049199; C030047H17Rik; D11Moh35; E2 ubiquitin-conjugating enzyme Z; HOYS7; RGD1308347; uba6-specific E2 conjugating enzyme 1; UBA6-specific enzyme E2; UBE2Z; Ubiquitin carrier protein Z; ubiquitin conjugating enzyme E2 Z; ubiquitin conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2 Z; ubiquitin-conjugating enzyme E2Z; ubiquitin-conjugating enzyme E2Z (putative); ubiquitin-protein ligase Z; Use1
Gene Symbols UBE2Z
Host Species Rabbit
Immunogen Recombinant Human Ubiquitin-conjugating enzyme E2 Z protein (115-354AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65264
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.