Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE3C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110540
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110540 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110540 Supplier Invitrogen™ Supplier No. PA5110540
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PISLEDLKSF TNYSGGYSAD HPVIKVFWRV VEGFTDEEKR KLLKFVTSCS RPPLLGFKEL YPAFCIHNGG SDLERLPTAS TCMNLLKLPE FYDETLLRSK LLYAIECAAG FELS Product note: Prior to December 2023, PA5-110540 was manufactured using a Recombinant fusion protein immunogen containing a sequence corresponding to amino acids 1-270 of human UBE3C. The affected lots are: WE3272737, WL3440149, XG3626363, 334B8A30, WH3338145, and VE2999302. Beginning in January 2024, all lots have been produced using a Recombinant fusion protein immunogen containing a sequence corresponding to amino acids 970-1083 of human UBE3C. Refer to the datasheets linked below for lot-specific immunogen information.

E3 ubiquitin-protein ligase that accepts ubiquitin from the E2 ubiquitin-conjugating enzyme UBE2D1 in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Can assemble unanchored poly-ubiquitin chains in either 'Lys-29'- or 'Lys-48'-linked polyubiquitin chains. Has preference for 'Lys-48' linkages. It can target itself for ubiquitination in vitro and may promote its own degradation in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE3C
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UBE3C
Gene Accession No. Q15386
Gene Alias AI853514; HectH2; HECT-type ubiquitin transferase E3C; KIAA0010; KIAA10; mKIAA0010; RGD1559986; Ube3c; ubiquitin protein ligase E3C; ubiquitin-protein isopeptide ligase (E3); ubiquitin-protein ligase E3C
Gene Symbols UBE3C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 970-1083 of human UBE3C (NP_0554862).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9690
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.