Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE3C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 123331AP150UL

Description
E3 ubiquitin-protein ligase that accepts ubiquitin from the E2 ubiquitin-conjugating enzyme UBE2D1 in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. Can assemble uchored poly-ubiquitin chains in either ′Lys-29′- or ′Lys-48′-linked polyubiquitin chains. Has preference for ′Lys-48′ linkages. It can target itself for ubiquitination in vitro and may promote its own degradation in vivo.Specifications
UBE3C | |
Polyclonal | |
Unconjugated | |
UBE3C | |
KIAA0010, KIAA10, UBE3C, ubiquitin protein ligase E3C | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100763, 362294, 9690 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15386, Q80U95 | |
Ube3c | |
UBE3C Fusion Protein Ag2966 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction