Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE4A Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298836MFV450
Description
UBE4A Polyclonal antibody specifically detects UBE4A in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
UBE4A | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
E4MGC133315, KIAA0126ubiquitination factor E4A (homologous to yeast UFD2), ubiquitination factor E4A (UFD2 homolog, yeast), UBOX2, UFD2ubiquitin conjugation factor E4 A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human UBE4A. | |
0.1 mL | |
Cell Biology | |
9354 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction