Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBE4A Antibody (SR1103), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32243450UL
Description
UBE4A Monoclonal antibody specifically detects UBE4A in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
UBE4A | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence | |
SR1103 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
E4MGC133315, KIAA0126ubiquitination factor E4A (homologous to yeast UFD2), ubiquitination factor E4A (UFD2 homolog, yeast), UBOX2, UFD2ubiquitin conjugation factor E4 A | |
A synthesized peptide derived from human UBE4A (Uniprot #: Q14139) | |
50 μg | |
Cell Biology | |
9354 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction