Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBE4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120865 Supplier Invitrogen™ Supplier No. PA5120865
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: MCF7, HepG2, 293T, Mouse kidney, Mouse pancreas, Mouse liver. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: FLLDEAIQYL SKIKIQQIEK DRGEWDSLTP EARREKEAGL QMFGQLARFH NIMSNETIGT LAFLTSEIKS LFVHPFLAER IISMLNYFLQ HLVGPKMGAL KVKDFSEFDF KPQQLVSDIC TIYLNLGDEE NFCATVPKDG RSYSPTLFAQ TVRVLKKINK PGNMIMAFSN LAERIKSLAD LQQQEEETYA DACDEFLDPI MSTLMCDPVV LPSSRVTVDR STIARHLLSD QTDPFNRSPL TMDQIRPNTE LKEKIQRWLA ERKQQKEQLE.

Ubiquitination requires a ubiquitin-activating enzyme, a ubiquitin-conjugating enzyme, and usually a substrate-specific ubiquitin-protein ligase. Multiubiquitination needs a fourth factor, to bind to the ubiquitin moieties of preformed conjugates and catalyze ubiquitin chain assembly in conjunction with the first three proteins. Human protein KIAA0126, with a predicted 1,073 amino acid residues, is a member of the novel protein family defined by ubiquitination conjugation factor E4. In yeast, E4 activity is linked to cell survival under stress conditions, indicating that eucaryotes use E4-dependent proteolysis pathways for multiple cellular functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBE4A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ube4a
Gene Accession No. E9Q735, Q14139, Q6P7A2
Gene Alias 4732444G18Rik; 9930123J21Rik; Ab2-232; E4; KIAA0126; RING-type E3 ubiquitin transferase E4 A; UBE4A; Ubiquitin conjugation factor E4 A; ubiquitination factor E4A; ubiquitination factor E4A (UFD2 homolog, yeast); ubiquitination factor E4A, UFD2 homolog; UBOX2; UFD2; UFD2b
Gene Symbols Ube4a
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140630, 315608, 9354
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.